Order Entry
Belgium
ContactUsLinkComponent
Pyridine, low water content ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Flammable
Corrosive
Irritant
Health
Environment
Pyridine, low water content ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Catalog # 9393-02
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Health
Environment
Pyridine, low water content ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Catalog # 9393-02
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    ≥ 99.0 %
  • Assay (by GC, corrected for water):
    ≥99.0 %
  • Ammonia (as NH₃):
    ≤0.002 %
  • Color (APHA):
    ≤10
  • Reducing Substances:
    Passes Test
  • Residue after Evaporation:
    ≤5 ppm
  • Solubility in H₂O:
    Passes Test
  • Sulfate (SO₄):
    ≤0.001 %
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 nm:
    ≤0.005
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 375 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 340 nm:
    ≤0.10
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 330 nm:
    ≤1.00
  • Water (by KF, volumetric):
    ≤0.01 %
  • Chloride (Cl):
    ≤5 ppm
  • Trace Impurities - Copper (Cu):
    ≤5 ppm

Specification Test Results

About this item

 :
Merck Index:13,08060
Boiling Pt:115 °C (1013 hPa)
Melting Pt:–44 °C
Density:0.983 g/cm³ (20 °C)
Flash Pt:17 °C
ADR:3,II
 :Packaged under nitrogen.
 :79.1 g/mol
EINECS:203-809-9
UN:1282
 :GHS02
 :GHS05
 :GHS07
 :GHS08
 :GHS09
Storage Temperature:Ambient
CAS Number:110-86-1
Formula:C₅H₅N
MDL Number:MFCD00011732