

Pyridine ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-86-1
:
Formula:C₅H₅N
MDL Number:MFCD00011732
Merck Index:14,7970
Boiling Pt:115 °C
Melting Pt:–42 °C
Density:0.978
Flash Pt:20 °C (68 °F)
UN:1282
ADR:3,II
:79.1 g/mol
EINECS:203-809-9
:GHS02
:GHS07
Storage Temperature:Ambient
Frequently Bought Together