Pyridine 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Azabenzene, Azine
- CAS Number:110-86-1
- Formula:C₅H₅N
- MDL Number:MFCD00011732
- Molecular Weight:79.1 g/mol
- Merck Index:13,08060
- Boiling Pt:115 °C (1013 hPa)
- Melting Pt:–42 °C
- Density:0.978 g/cm³
- Flash Pt:17 °C
- Storage Temperature:Ambient
- EINECS:203-809-9
- UN:1282
- ADR:3,II
3 Options Available Below
Product Details & Documents
Synonyms: Azabenzene, Azine
- CAS Number:110-86-1
- Formula:C₅H₅N
- MDL Number:MFCD00011732
- Molecular Weight:79.1 g/mol
- Merck Index:13,08060
- Boiling Pt:115 °C (1013 hPa)
- Melting Pt:–42 °C
- Density:0.978 g/cm³
- Flash Pt:17 °C
- Storage Temperature:Ambient
- EINECS:203-809-9
- UN:1282
- ADR:3,II
Documents
ACRO38656
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.05 %
- GC:≥99.0 %
- UV:at 310 nm A: ≤1
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



