Order Entry
Belgium
ContactUsLinkComponent
Tetrahydrofuran stabilised, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Flammable
Health
Irritant
Tetrahydrofuran stabilised, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Flammable
Health
Irritant
Tetrahydrofuran stabilised, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -108.5 °C
  • Molar Mass:
    72.11 g/mol
  • Boiling Point:
    65 - 66 °C
  • Vapor Pressure:
    173 hPa (20 °C)
  • Flash Point:
    -21.5 °C
  • Refractive Index:
    1.407 (20 °C)
  • Explosion Limit:
    1.5 - 12.4 %(V)
  • Density:
    0.89 g/cm3 (20 °C)
  • pH:
    7 - 8 (200 g/l, H2O, 20 °C)
  • Ignition Point:
    215 °C
  • Evaporation number:
    2.3

Specification Test Results

Related Information

Frequently Bought Together