Order Entry
Belgium
Orders LinkContactUsLinkComponent
Infrared printer

Infrared printer

Infrared printer Ir®DA with thermal paper and four batteries, 147×77×47 mm, 430 g
Catalog # 620-1903
Supplier:   TESTO
Infrared printer
Catalog # 620-1903
Supplier:  TESTO
Infrared printer
Infrared printer

Infrared printer Ir®DA with thermal paper and four batteries, 147×77×47 mm, 430 g

Price
261,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The universal IR printer completes data transfers in two seconds. This printer allows users to evaluate restoration measures of air moisture, material equilibrium moisture, and pressure dew point quickly and without error.

  • Hands-free operation
  • Wireless infrared interface for fast data transfer
  • Provides fast, easy to read printouts of measurements
  • Energy saving through auto-off feature
  • Instrument memory for up to 10000 readings

Delivery information: Printer supplied wih a wireless infrared interface, one roll of thermal paper, and four AA batteries.

Specifications
  • Description:
    Infrared printer Ir®DA with thermal paper and four batteries, 147×77×47 mm, 430 g
  • Seq:
    01
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Enhanced thermal paper
    Enhanced thermal paper
    TEST0554.0568
    Log in for availability
    1 * 6 ST - 29,00 €

Product Information

  • Catalog #
    Availability
    Price
  • International mains unit, 2000 mA, for printer
    International mains unit, 2000 mA, for printer
    620-1593
    Log in for availability
    1 * 1 ST - 53,00 €