Order Entry
Belgium
ContactUsLinkComponent
Accessories for Moxi™ Z
Accessories for Moxi™ Z
Supplier:  Orflo
Certificates
undefined
Accessories for Moxi™ Z
Supplier:  Orflo

Specifications

  • Catalog No:
  • 701-0252
  • ORFLMXC012
  • ORFLMXC011
  • Model:
  • Type S cassettes
  • Type M cassettes
  • Concentration:
  • 3000 – 1750000 cells/ml
  • 3000 – 500000 cells/ml
  • Cell size:
  • 14 - 9202 fL
  • 34 - 20580 fL
  • Sample volume:
  • 75 µl
  • 75 µl
  • Dynamic range:
  • 3 - 26 µm
  • 4 - 34 µm
  • Measurement time:
  • 15 s
  • 8 s

Specifications

Frequently Bought Together