Order Entry
Belgium
Orders LinkContactUsLinkComponent
Zinc oxide light ≥98%, TECHNICAL

Zinc oxide light ≥98%, TECHNICAL

Plastic bottle for solids, 1 kg
Catalog # 29199.295
Environment
Supplier:   VWR Chemicals
Zinc oxide light ≥98%, TECHNICAL
Catalog # 29199.295
Supplier:  VWR Chemicals
CAS Number:  1314-13-2
Zinc oxide light ≥98%, TECHNICAL
Zinc oxide light ≥98%, TECHNICAL

Plastic bottle for solids, 1 kg

CAS Number:  1314-13-2
Price
122,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Zinci oxidum, Zinc (II) oxide
  • CAS Number:1314-13-2
  • Formula:ZnO
  • MDL Number:MFCD00011300
  • Molecular Weight:81.39 g/mol
  • Merck Index:12,10279
  • Boiling Pt:2360 °C (1013 hPa)
  • Melting Pt:1975 °C
  • Density:5.6 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-222-5
  • UN:3077
  • ADR:9,III
Specifications

Specification Test Results

  • Assay:
    Min. 98 %

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.