Order Entry
Belgium
ContactUsLinkComponent
6-Nitro-1-indanone
6-Nitro-1-indanone
Catalog # MOLE12551217-1G
Supplier:  Molekula
CAS Number:  
6-Nitro-1-indanone
Catalog # MOLE12551217-1G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :177.16 g/mol
EINECS:246-366-7
CAS Number:24623-24-3
Formula:C₉H₇NO₃
MDL Number:MFCD06656903