Phosphoryl trichloride
VWRS1534.1000
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
:
:153.33 g/mol
:12,07506
:107 °C (1013 hPa)
:1 °C
:1.645 g/cm³ (20 °C)
:Ambient
:233-046-7
:1810
:8,I
:GHS06
:GHS05
:GHS08
:10025-87-3
:POCl₃
:MFCD00011443