Phosphoryl trichloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10025-87-3
:
Formula:POCl₃
MDL Number:MFCD00011443
:153.33 g/mol
Merck Index:12,07506
Boiling Pt:107 °C (1013 hPa)
Melting Pt:1 °C
Density:1.645 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1810
ADR:6.1 (8),I
:M-Clarity™ quality level = MQ200
EINECS:233-046-7
:GHS06
:GHS05
:GHS08
Limited Time Sale
Select items below are currently on sale
Related Information
Certificates
Search by Lot NumberMERC8.22339.1000_TDS_EN ProductPage_822339 ProductPage_822339
Product specification
Merck Individual products and tailor-made chemicals for your technical applications
Product Literature
msds_822339
SDS
Frequently Bought Together