Phosphoryl trichloride 99%
:10025-87-3
:
:POCl₃
:MFCD00011443
:153.33 g/mol
:12,07506
:107 °C (1013 hPa)
:1.2 °C
:1.645 g/cm³
:Ambient
:233-046-7
:1810
:6.1,I
:GHS06
:GHS08
:GHS05
- Appearance:Clear liquid
- Refractive index:1.4600 to 1.4620 (20°C, 589 nm)
- Titration Argentometric:≥98.5 % (ex Cl)
- Color scale:≤40 APHA
- Phosphorus trichloride (PCl3):≤0.3 %
Frequently Bought Together