Silica gel, Pore size: 90 Å 0.035-0.070 mm for chromatography
:7631-86-9
:112926-00-8
:SiO₂
:MFCD00011232
:60.08 g/mol
:2230 °C (1013 hPa)
:1710 °C
:Ambient
:01-2119379499-16
- Appearance:White Powder
- Particle size:35 to 70 µm
- Bulk density:385 to 440 g/l
- pH:6.5 to 7.5 (5 % w/w suspension)
- Residual water:2 to 4 % (160°C)
Frequently Bought Together