Order Entry
Belgium
ContactUsLinkComponent
Orthoboric acid, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Health
Orthoboric acid, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Health
Orthoboric acid, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    46.5 g/l (20 °C)
  • Melting Point:
    171 °C (decomposition)
  • Molar Mass:
    61.83 g/mol
  • Bulk Density:
    400 - 600 kg/m3
  • Vapor Pressure:
    2.7 hPa (20 °C)
  • Density:
    1.44 g/cm3 (20 °C)
  • pH:
    3.8 - 4.8 (33 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together