Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

D-(+)-Glucose

500 mg
Catalog # 47249.
Supplier:   Merck
D-(+)-Glucose
Catalog # 47249.
Supplier:  Merck
CAS Number:  50-99-7
D-(+)-Glucose
D-(+)-Glucose

500 mg

CAS Number:  50-99-7
Price
45,50 €
Log in for availability
1 * 500 mg
Product Details & Documents
Synonyms:  Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
  • CAS Number:50-99-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148912
  • Molecular Weight:180.16 g/mol
  • Merck Index:14,04459
  • Melting Pt:146 °C
  • Density:1.54 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-075-1

 

Specifications

Specifications

  • Pk:
    500 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.