Order Entry
Belgium
ContactUsLinkComponent
Gallium(III) chloride, anhydrous ≥99.999%, granular, PURATREM
Corrosive
Gallium(III) chloride, anhydrous ≥99.999%, granular, PURATREM
Catalog # STRE93-3140.0025
CAS Number:  
Corrosive
Gallium(III) chloride, anhydrous ≥99.999%, granular, PURATREM
Catalog # STRE93-3140.0025
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :176.08 g/mol
Boiling Pt:201.3 °C
Melting Pt:77.9 °C
Density:2.47
EINECS:236-610-0
UN:1759
ADR:8,II
 :GHS05
CAS Number:13450-90-3
Formula:Cl₃Ga
MDL Number:MFCD00011018