Orders LinkContactUsLinkComponent
Glass beads
Glass beads
Glass beads
Supplier:  MARIENFELD
Certificates
Glass beads
6 Options Available Below
Product Details & Documents
Made of soda lime glass.
  • Solid
  • Polished
Glass beads are used for mixing or milling. The smaller the beads and the higher their quantity the more intense is the mixing or milling.
Packaging: In poly bag of 500 g or 1000 g each.
Documents
Product Family Options
Product Information
  • Ø
    Availability
    Price
  • MARI4901001
    Glass beads, solid, 1000 g pack
    Glass beads, solid, 1000 g pack
    1 mm
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 46,90 €
    46,90 €
    1 * 1 kg
       46,90 €
  • MARI4901002
    Glass beads, solid, 500 g pack
    Glass beads, solid, 500 g pack
    2 mm
    Log in for availability
    Log in for availability
    1 * 0,5 kg
    1 * 0,5 kg - 18,80 €
    18,80 €
    1 * 0,5 kg
       18,80 €
  • MARI4901003
    Glass beads, solid, 500 g pack
    Glass beads, solid, 500 g pack
    3 mm
    Log in for availability
    Log in for availability
    1 * 0,5 kg
    1 * 0,5 kg - 18,80 €
    18,80 €
    1 * 0,5 kg
       18,80 €
  • MARI4901004
    Glass beads, solid, 500 g pack
    Glass beads, solid, 500 g pack
    4 mm
    Log in for availability
    Log in for availability
    1 * 0,5 kg
    1 * 0,5 kg - 18,80 €
    18,80 €
    1 * 0,5 kg
       18,80 €
  • MARI4901005
    Glass beads, solid, 500 g pack
    Glass beads, solid, 500 g pack
    5 mm
    Log in for availability
    Log in for availability
    1 * 0,5 kg
    1 * 0,5 kg - 18,80 €
    18,80 €
    1 * 0,5 kg
       18,80 €
  • MARI4901006
    Glass beads, solid, 1000 g pack
    Glass beads, solid, 1000 g pack
    6 mm
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 38,80 €
    38,80 €
    1 * 1 kg
       38,80 €