Bed pan
2500 ml
KART586
: KARTELL
Bed pan moulded in white PP.
- Withstand temperatures up to 120 °C
- Very good chemical resistance
- Easy to clean
- Capacity:2500 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



