Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Drip pans
Drip pans
Supplier:   USBECK Laborgeräte
Drip pans
Certificates
Drip pans
2 Options Available Below
Product Details & Documents
Made of stainless steel.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean
With removable gridiron.
Product Family Options
Product Information
  • Largeur
    Hauteur
    Longueur
    Availability
    Price
  • 200 mm
    27 mm
    320 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 143,00 €
    143,00 €
    1 * 1 ST
       143,00 €
  • 270 mm
    27 mm
    440 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 191,00 €
    191,00 €
    1 * 1 ST
       191,00 €