Order Entry
Belgium
ContactUsLinkComponent
 
Magnesium chloride, anhydrous ≥98%, TECHNICAL
Catalog # 26123DC
Supplier:  VWR Chemicals
CAS Number:  
undefined
Magnesium chloride, anhydrous ≥98%, TECHNICAL
Catalog # 26123DC
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Assay:
    Min. 98 %

Specification Test Results

About this item

 :
 :95.21 g/mol
Merck Index:13,05684
Boiling Pt:1412 °C (1013 hPa)
Melting Pt:715 °C
Density:2.32 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7786-30-3
Formula:MgCl₂
MDL Number:MFCD00011106