Capillary columns, BP10
These columns are suitable for environmental analyses (EPA methods 608 and 8081), pesticides/herbicides, drugs of abuse, pharmaceuticals.
14% cyanopropylphenyl polysiloxane
Max. temperature: 0,22 - 0,32 mm ID -20 °C to 280/300 °C ; 0,53 mm ID -20 °C to 260/280 °C
Suitable replacement for: DB-1701, Rtx-1701, SPB-7, HP-1701, CP-Sil 19CB, 007-1701, PE-1701, SP-1701
Frequently Bought Together