Order Entry
Belgium
ContactUsLinkComponent
Sulphuric acid 96% ≥96%, Ultrapure for trace analysis, Supelco®
Sulphuric acid 96% ≥96%, Ultrapure for trace analysis, Supelco®
Catalog # 1.01516.0250
Supplier:  Merck
CAS Number:  
Sulphuric acid 96% ≥96%, Ultrapure for trace analysis, Supelco®
Catalog # 1.01516.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 mL
  • Seq:
    2590
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble,(caution ! development of heat)
  • Melting Point:
    -20 °C
  • Molar Mass:
    98.08 g/mol
  • Boiling Point:
    335 °C
  • Vapor Pressure:
    0.0001 hPa (20 °C)
  • Density:
    1.84 g/cm3 (20 °C)
  • pH:
    0.3 (49 g/l, H2O, 25 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

About this item

 :
 :98.08 g/mol
Boiling Pt:∼335 °C (1013 hPa)
EINECS:231-639-5
UN:1830
 :GHS05
Storage Temperature:Ambient
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589
Melting Pt:10.38
Density:1.84 g/cm³ (20 °C)
ADR:8 II
 :M-Clarity™ quality level = MQ100