Sulphuric acid 96% ≥96%, Suprapur® for trace analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
Boiling Pt:∼335 °C (1013 hPa)
Melting Pt:10.38
Density:1.84 g/cm³ (20 °C)
Molecular Weight:98.08 g/mol
EINECS:231-639-5
ADR:8 II
:M-Clarity™ quality level = MQ100
UN:1830
:GHS05
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) soluble,(caution ! development of heat)
- Melting Point:-20 °C
- Molar Mass:98.08 g/mol
- Boiling Point:335 °C
- Vapor Pressure:0.0001 hPa (20 °C)
- Density:1.84 g/cm3 (20 °C)
- pH:0.3 (49 g/l, H2O, 25 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Certificates
Search by Lot NumberMerck Pharma Segment Brochure Merck Ultrapur and Suprapur® High Purity Acids and Bases
Product Literature
msds_100714
SDS
ProductPage_100714
Product specification
Frequently Bought Together