
Sulphuric acid 95-97%, EMSURE® ISO for analysis, Supelco®
: Merck
:7664-93-9
:
:H₂SO₄
:MFCD00064589
:98.08 g/mol
:14,08974
:∼335 °C (1013 hPa)
:3 °C
:1.84 g/cm³ (20 °C)
:1830
:8,II
:M-Clarity™ quality level = MQ300
:231-639-5
:GHS05
:Ambient
- Solubility:(20 °C) soluble,(caution ! development of heat)
- Melting Point:-20 °C
- Molar Mass:98.08 g/mol
- Boiling Point:335 °C
- Vapor Pressure:0.0001 hPa (20 °C)
- Density:1.84 g/cm3 (20 °C)
- pH:0.3 (49 g/l, H2O, 25 °C)
Frequently Bought Together