Order Entry
Belgium
ContactUsLinkComponent
Sodium sulphate, anhydrous ≥99.0% (by titrimetric analysis), Purum p.a.
Sodium sulphate, anhydrous ≥99.0% (by titrimetric analysis), Purum p.a.
Certificates
undefined
Sodium sulphate, anhydrous ≥99.0% (by titrimetric analysis), Purum p.a.
Certificates

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Loss on drying (130°C):
    Max. 1 %
  • pH (5 %, 20°C):
    5.0 - 7.5
  • Calcium (Ca):
    Max. 0.005 %
  • Cadmium (Cd):
    Max. 0.005 %
  • Cobalt (Co):
    Max. 0.005 %
  • Copper (Cu):
    Max. 0.005 %
  • Iron (Fe):
    Max. 0.005 %
  • Potassium (K):
    Max. 0.01 %
  • Nickel (Ni):
    Max. 0.005 %
  • Lead (Pb):
    Max. 0.005 %
  • Zinc (Zn):
    Max. 0.005 %
  • Chloride (Cl):
    Max. 0.02 %

Specification Test Results

Related Information

Frequently Bought Together