Order Entry
Belgium
ContactUsLinkComponent
Sodium sulphate ≥99.0%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Sodium sulphate ≥99.0%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Sodium sulphate ≥99.0%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    min. 99.0 %
  • Calcium, Magnesium and R2O3 Precipitate:
    max. 0.01 %
  • Chloride (Cl):
    max. 0.001 %
  • Insoluble Matter:
    max. 0.01 %
  • Potassium (K) (by AAS):
    max. 0.01 %
  • Loss on Ignition:
    max. 0.5 %
  • pH of 5% Solution at 25°C:
    5.2 - 9.2
  • Arsenic (As):
    max. 0.5
  • Iron (Fe):
    max. 5
  • Heavy Metals (as Pb):
    max. 5
  • Nitrogen Compounds (as N):
    max. 5

Specification Test Results

Related Information

Frequently Bought Together