

Sodium metabisulphite 97+% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7681-57-4
:
Formula:Na₂S₂O₅
MDL Number:MFCD00167602
:190.11 g/mol
Merck Index:12,08784
Melting Pt:150 °C (dec.)
Density:1,48 g/cm³ (20 °C)
EINECS:231-673-0
REACH:01-2119531326-45
:GHS07
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Titration Iodimetric:≥97.0 %
- Chloride (Cl):≤500 ppm
- Thiosulphate (S2O3):≤500 ppm
- Iron (Fe):≤20 ppm
- Heavy metals (as Pb):≤10 ppm
- Insoluble matter:≤0.005 %
Specification Test Results
Frequently Bought Together