



Lithium sulphide ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12136-58-2
:
Formula:Li₂S
MDL Number:MFCD00011085
:45.95 g/mol
Boiling Pt:1527 °C (1013 hPa)
Melting Pt:900…975 °C
Density:1.66
EINECS:235-228-1
UN:3134
ADR:4.3,II
:GHS02
:GHS06
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together