Disodium hydrogen phosphate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7558-79-4
:
Formula:Na₂HPO₄
MDL Number:MFCD00003496
:141.96 g/mol
Merck Index:12,08805
Melting Pt:250 °C
Density:1.53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-448-7
REACH:01-2119489797-11
Specification Test Results
- Assay:Min. 99 %
- Heavy metals (as Pb):Max. 10 ppm
- Cl (Chloride):Max. 0.02 %
- SO4 (Sulphate):Max. 0.05 %
- As (Arsenic):Max. 2 ppm
- Fe (Iron):Max. 50 ppm
Specification Test Results
Frequently Bought Together