Order Entry
Belgium
ContactUsLinkComponent
Disodium hydrogen phosphate
Disodium hydrogen phosphate
Catalog # MOLE40775320
Supplier:  Molekula
CAS Number:  
Disodium hydrogen phosphate
Catalog # MOLE40775320
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :141.96 g/mol
Merck Index:12,08805
Melting Pt:250 °C
Density:1.53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-448-7
REACH:01-2119489797-11
CAS Number:7558-79-4
Formula:Na₂HPO₄
MDL Number:MFCD00003496