Order Entry
Belgium
ContactUsLinkComponent
Disodium hydrogen phosphate, anhydrous ≥99.0%, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Disodium hydrogen phosphate, anhydrous ≥99.0%, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Disodium hydrogen phosphate, anhydrous ≥99.0%, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Assay (alkalimetric):
    ≥99.0%
  • Insoluble matter:
    ≤0.01%
  • pH-value (5%; water, 25 °C):
    8.7 - 9.3
  • Chloride (Cl):
    ≤0.002%
  • Sulfate (SO₄):
    ≤0.005%
  • Heavy metals (as Pb):
    ≤0.001%
  • Cu (Copper):
    ≤0.0003%
  • Fe (Iron):
    ≤0.001%
  • K (Potassium):
    ≤0.01%
  • Pb (Lead):
    ≤0.001%
  • Loss on Drying (105 °C):
    ≤0.2%

Specification Test Results

Related Information

Frequently Bought Together