Order Entry
Belgium
ContactUsLinkComponent
 
 
Orthophosphoric acid ≥99.0% (by titrimetric analysis), crystallised, Puriss. p.a., Fluka™
 
 
Orthophosphoric acid ≥99.0% (by titrimetric analysis), crystallised, Puriss. p.a., Fluka™
 

 

  • Assay:
    99.0 - 101.0 %
  • Melting point:
    38 - 42 °C
  • Arsenic (As):
    Max. 1 ppm
  • Calcium (Ca):
    Max. 10 ppm
  • Cadmium (Cd):
    Max. 1 ppm
  • Cobalt (Co):
    Max. 1 ppm
  • Chromium (Cr):
    Max. 1 ppm
  • Copper (Cu):
    Max. 1 ppm
  • Iron (Fe):
    Max. 10 ppm
  • Potassium (K):
    Max. 5 ppm
  • Magnesium (Mg):
    Max. 1 ppm
  • Manganese (Mn):
    Max. 1 ppm
  • Sodium (Na):
    Max. 50 ppm
  • Nickel (Ni):
    Max. 1 ppm
  • Lead (Pb):
    Max. 1 ppm
  • Zinc (Zn):
    Max. 1 ppm
  • Chloride (Cl):
    Max. 5 ppm
  • Nitrate (NO₃):
    Max. 5 ppm
  • Sulfate (SO₄):
    Max. 30 ppm
  • Phosphite,Hypoph. (H₃PO₃):
    Max. 500 ppm
  • KMnO₄ reducing matter (as O):
    Max. 20 ppm
  • Vapor Pressure:
    5 mmHg ( 25 °C)

 

 

Frequently Bought Together