
Sodium hydroxide 97.0-100.5%, pellets NF, JP, Ph. Eur., BP
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Merck Index:14,08627
Boiling Pt:1390 °C (1013 hPa)
Melting Pt:323 °C
Density:2.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1823
ADR:8,II
REACH:01-2119457892-27
:GHS05
Specification Test Results
- Appearance:Conforms (see CoA/CoS)
- Heavy metals (as Pb):Max. 20 ppm
- Residual solvents:Passes test
- As (Arsenic):Max. 1 ppm
- Cd (Cadmium):Max. 1 ppm
- Co (Cobalt):Max. 1 ppm
- Hg (Mercury):Max. 1 ppm
- Ni (Nickel):Max. 1 ppm
- Pb (Lead):Max. 5 ppm
- V (Vanadium):Max. 1 ppm
- Conforms to Ph.Eur./NF/BP/JP:Passes test
- Ph.Eur./BP specifications::Min. 11.0
- Assay (calculated as NaOH) [Ph.Eur./BP]:97.0 --> 100.5 %
- Identification A [Ph.Eur./BP]:Min. 11.0
- Identification B:Passes test Ph.Eur./BP
- Appearance of solution:Passes test Ph.Eur./BP
- CO3 (as Na2CO3) [Ph.Eur./BP]:Max. 2.0 %
- Cl (Chloride) [Ph.Eur./BP]:Max. 200 ppm
- SO4 (Sulphate) [Ph.Eur./BP]:Max. 200 ppm
- Fe (Iron) [Ph.Eur./BP]:Max. 10 ppm
- NF specifications::Max. 1 ppm
- Assay (total alkali) [NF]:95.0 --> 100.5 %
- Sodium carbonate [NF]:Max. 3.0 %
- Content of sodium [NF]:54.0 --> 59.8 %
- Identification A:Passes test NF
- K (Potassium) [NF]:Max. 0.5 %
- Insoluble substances and organic matter:Passes test NF
- Assay (calculated as NaOH) [JP]:Min. 95.0 %
- Identification 1:Passes test JP
- Identification 2:Passes test JP
- Clarity and colour of solution:Passes test JP
- Cl (Chloride) [JP]:Max. 0.050 %
- K (Potassium):Passes test JP
- Sodium carbonate [JP]:Max. 2.0 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 6L Plastic bottle for solids DIN 80 (ZPACK13256000)
Packaging Information
28248_SODIUM HYDROXIDE PELLETS PH.EUR_ALLERGEN_REV2 Allergen_VWRC_28248_SODIUM HYDROXIDE PELLETS PH.EUR.pdf
ALLERGEN
28248_SODIUM HYDROXIDE PELLETS. PH.EUR_BSE_REV2
BSE/TSE Certificates
28248_SODIUM HYDROXIDE PELLETS. PH.EUR_LATEX_REV1 28248_SODIUM HYDROXIDE PELLETS. PH.EUR_MELAMINE_REV1 28248_SODIUM HYDROXIDE PELLETS. PH.EUR_PESTICIDES_REV0 28248_SODIUM HYDROXIDE PELLETS. PH.EUR_PHTALATE_REV1 Sodium Hydroxide 97 - 100,5%, Pellets Ph. Eur., NF, BP, JP
Additional Product Details
spec_VWRC28248.298 spec_VWRC28248.298 spec_VWRC28248.367 spec_VWRC28248.367 spec_VWRC28248.460
Product specification
VWR Chemicals for Biopharma and Pharma Production
Product Literature
Frequently Bought Together