Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sodium hydroxide

Sodium hydroxide

Plastic bottle for solids, 1 kg
Catalog # MOLEM80406338
Corrosive
Marketsource
Supplier:   Molekula
Sodium hydroxide
Catalog # MOLEM80406338
Supplier:  Molekula
Marketsource
CAS Number:  1310-73-2
Sodium hydroxide

Plastic bottle for solids, 1 kg

CAS Number:  1310-73-2
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,08627
  • Boiling Pt:1390 °C (1013 hPa)
  • Melting Pt:323 °C
  • Density:2.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1823
  • ADR:8,II
  • REACH:01-2119457892-27
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.