About this item
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Boiling Pt:1390 °C (1013 hPa)
- Melting Pt:323 °C
- Density:2.13 g/cm³ (20 °C)
- EINECS:215-185-5
- ADR:8,II
- :M-Clarity™ quality level = MQ300
- Storage Temperature:Ambient
- UN:1823
- REACH:01-2119457892-27
5 Options Available Below
Product Details & Documents
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Boiling Pt:1390 °C (1013 hPa)
- Melting Pt:323 °C
- Density:2.13 g/cm³ (20 °C)
- EINECS:215-185-5
- ADR:8,II
- :M-Clarity™ quality level = MQ300
- Storage Temperature:Ambient
- UN:1823
- REACH:01-2119457892-27
Documents
Certificates
Search by Lot NumberMERC1.06498.0500_TDS_EN MERC1.06498.1000_TDS_EN MERC1.06498.5000_TDS_EN ProductPage_106498 Spec MERC1.06498.1000.pdf
Product specification
msds_106498
SDS
Specifications for Product Family
Specification Test Results
- Assay (acidimetric, NaOH):99% min
- Carbonate (as Na₂CO₃):1.0% max
- Chloride (Cl):0.012% max
- Phosphate (PO₄):0.0005% max
- Silicate (SiO₂):0.001% max
- Sulfate (SO₄):0.010% max
- Total nitrogen (N):0.0003% max
- Heavy metals (as Pb):0.0005% max
- Al (Aluminium):0.0005% max
- As (Aresnic):0.0001% max
- Ca (Calcium):0.0005% max
- Cu (Copper):0.0002% max
- Fe (Iron):0.0005% max
- K (Pottassium):0.05% max
- Mg (Magnesium):0.0005% max
- Ni (Nickel):0.00025% max
- Pb (Lead):0.0005% max
- Zn (Zinc):0.001% max
- Zinc (Zn):0.001% max
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



