Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Corrosive
Sodium hydroxide, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
Corrosive
Sodium hydroxide, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    min. 98 %
  • Carbonate (as Na2CO3):
    max. 1 %
  • Potassium (K):
    max. 0.05 %
  • Silica (SiO2):
    max. 0.001 %
  • Trace Impurities (in ppm)Aluminium (Al):
    max. 5
  • Trace Impurities (in ppm)Calcium (Ca):
    max. 5
  • Trace Impurities (in ppm)Chloride (Cl):
    max. 10
  • Trace Impurities (in ppm)Iron (Fe):
    max. 5
  • Trace Impurities (in ppm)Nickel (Ni):
    max. 5
  • Trace Impurities (in ppm)Heavy Metals (as Pb):
    max. 5
  • Trace Impurities (in ppm)Phosphate (PO4):
    max. 5
  • Trace Impurities (in ppm)Sulfate (SO4):
    max. 5
  • Trace Impurities (in ppm)Total Nitrogen (N):
    max. 3

Specification Test Results

Related Information

Frequently Bought Together