
Sodium hydroxide ≥97%, pellets, Proteomics Grade
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Merck Index:14,08627
Boiling Pt:1390 °C (1013 hPa)
Melting Pt:318 °C
Density:2.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1823
ADR:8,II
REACH:01-2119457892-27
:GHS05
Specification Test Results
- Chloride:<0.005%
- DNase:none detected
- Heavy Metals:<0.002 %
- Iron:≤0.001%
- Mercury:≤0.1 ppm
- Nickel:≤0.001%
- Nitrogen Compounds:≤0.001%
- Phosphate:≤0.001%
- Potassium:≤0.02%
- Protease:none
- Purity:≥97%
- RNase:none detected
- Sodium Carbonate:≤1%
- Sulphate:≤0.003%
Specification Test Results
Frequently Bought Together