Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Lithium hydroxide, Supelco®

Corrosive
Irritant
Supplier:   Merck
Lithium hydroxide, Supelco®
Supplier:  Merck
Certificates

2 Options Available Below

Product Details & Documents
  • CAS Number:1310-65-2
  • Formula:LiOH
  • MDL Number:MFCD00011095
  • Molecular Weight:23.95 g/mol
  • Merck Index:13,05556
  • Boiling Pt:924 °C (1013 hPa)
  • Melting Pt:462 °C
  • Density:1.46 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-183-4
  • ADR:8,II
  •  :M-Clarity™ quality level = MQ300
  • UN:2680
Specifications for Product Family

Specification Test Results

  • Solubility:
    71 g/l (20 °C)
  • Melting Point:
    462 °C
  • Molar Mass:
    23.95 g/mol
  • Bulk Density:
    550 kg/m3
  • Density:
    2.54 g/cm3
  • pH:
    12 (50 g/l, H2O, 50 °C)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 247,00 €
    247,00 €
    1 * 100 g
       247,00 €

  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 1 430,00 €
    1 430,00 €
    1 * 1 kg
       1 430,00 €