Lithium hydroxide
100
Catalog # 705-0056
Supplier: Hach
Jump to:
Product Details & Documents
- CAS Number:1310-65-2
- Formula:LiOH
- MDL Number:MFCD00011095
- Molecular Weight:23.95 g/mol
- Merck Index:13,05556
- Boiling Pt:924 °C (1013 hPa)
- Melting Pt:462 °C
- Density:1.46 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-183-4
- UN:2680
- ADR:8,II
Specifications
Specifications
- Pk:100



