Lithium hydroxide, anhydrous 98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 1310-65-2 Molecular Formula: LiOH Formula Weight: 23.95 Physical Form: -4+14 Mesh Density: 2.54 Appearance: White Odour: Odourless pH: 14 Melting Point: 462° Boiling Point: 924° Solubility: Soluble in water. Slightly soluble in alcohol Merck Reference: 14,5534 UN No.: 2680 MDL No.: MFCD00011095
- CAS Number:1310-65-2
- Formula:LiOH
- MDL Number:MFCD00011095
- Molecular Weight:23.95 g/mol
- Merck Index:14,5534
- Boiling Pt:924 °C
- Melting Pt:462 °C
- Density:2.54
- Storage Temperature:Ambient
- EINECS:215-183-4
- UN:2680
- ADR:8,II
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 1310-65-2 Molecular Formula: LiOH Formula Weight: 23.95 Physical Form: -4+14 Mesh Density: 2.54 Appearance: White Odour: Odourless pH: 14 Melting Point: 462° Boiling Point: 924° Solubility: Soluble in water. Slightly soluble in alcohol Merck Reference: 14,5534 UN No.: 2680 MDL No.: MFCD00011095
- CAS Number:1310-65-2
- Formula:LiOH
- MDL Number:MFCD00011095
- Molecular Weight:23.95 g/mol
- Merck Index:14,5534
- Boiling Pt:924 °C
- Melting Pt:462 °C
- Density:2.54
- Storage Temperature:Ambient
- EINECS:215-183-4
- UN:2680
- ADR:8,II



