Order Entry
Belgium
ContactUsLinkComponent
Syringe filters, Econofil™, Chromacol™
Syringe filters, Econofil™, Chromacol™
Catalog # 516-0653
Supplier:  CHROMACOL
undefined
Syringe filters, Econofil™, Chromacol™
Catalog # 516-0653
Supplier:  CHROMACOL

Specifications

  • Ø:
    4 mm
  • Membrane:
    CA
  • Pore size:
    0,20 µm
  • Seq:
    01
  • Pk:
    100

Specifications

About this item

The sample distribution rings promote uniform application of the sample across the membrane area. This feature maximises the available filltration area speed and reduces backpressure when filtering highly particulate samples.

  • Sample distribution rings distribute sample for maximum speed and uniform filtration across membrane surface
  • Membrane type and porosity clearly identified on each filter
  • Chemically resistant PP housing minimises extractables