Orders LinkContactUsLinkComponent
Counting chamber, McMaster
Counting chamber, McMaster
Supplier:   MARIENFELD
Counting chamber, McMaster
Supplier:  MARIENFELD
Certificates
Counting chamber, McMaster
Counting chamber, McMaster
2 Options Available Below
Product Details & Documents
This cell counter is suitable for liquid media.
  • Three counting grids and spare cover glass
  • Pair of clamps to fix the cover glass
  • Ground edges and rounded corners
Product Family Options
Cover glass
Product Information
  • Depth of chamber
    W×D
    Availability
    Price
  • MARI0335001
    Cover glass with three counting grids (10×10 mm) divided into 10 parts
    Cover glass with three counting grids (10×10 mm) divided into 10 parts
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 184,00 €
    184,00 €
    1 * 1 ST
       184,00 €
Product Information
  • Depth of chamber
    W×D
    Availability
    Price
  • 631-0695
    Counting chamber
    Counting chamber
    1,5 mm
    127×26 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 431,00 €
    431,00 €
    1 * 1 ST
       431,00 €