Orders LinkContactUsLinkComponent
Counting chambers, Howard
Counting chambers, Howard
Supplier:   MARIENFELD
Counting chambers, Howard
Supplier:  MARIENFELD
Certificates
Counting chambers, Howard
2 Options Available Below
Product Details & Documents
Counting chambers designed for the investigation of fruit juices.
Ø ring = 18 - 22 mm
Product Family Options
Product Information
  • Depth of chamber
    Availability
    Price
  • MARI0610940
    Counting chamber with double lines
    Counting chamber with double lines
    0,10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 264,00 €
    264,00 €
    1 * 1 ST
       264,00 €
  • MARI0611040
    Counting chamber
    Counting chamber
    0,10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 904,00 €
    904,00 €
    1 * 1 ST
       904,00 €
Accessories
Product Information
  • Catalog #
    L×W
    Thickness
    Availability
    Price
  • Cover glass with blue mask – 5×5 wells of Ø 1,382 mm
    Cover glass with blue mask – 5×5 wells of Ø 1,382 mm
    MARI0334001
    33×33 mm
    1,0 mm
    Log in for availability
    1 * 1 ST - 276,00 €
Product Information
  • Catalog #
    L×W
    Thickness
    Availability
    Price
  • Cover glass with ground edges
    Cover glass with ground edges
    MARI0333001
    25×40 mm
    0,4 mm
    Log in for availability
    1 * 1 ST - 6,10 €
Product Information
  • Catalog #
    L×W
    Thickness
    Availability
    Price
  • Cover glass with metal mask - 5×5 wells of Ø = 1,382 mm
    Cover glass with metal mask - 5×5 wells of Ø = 1,382 mm
    MARI0334002
    25×40 mm
    0,4 mm
    Log in for availability
    1 * 1 ST - 210,00 €