Counting chambers, Neubauer
Supplier: MARIENFELD
Jump to:
About this item
These counting chambers are designed for counting leucocytes and erythrocytes.
- Double net ruling
- Two counting grids with one division
: IVD 98/79/EC
: Supplied with two cover glasses.
4 Options Available Below
Product Details & Documents
These counting chambers are designed for counting leucocytes and erythrocytes.
- Double net ruling
- Two counting grids with one division
: IVD 98/79/EC
: Supplied with two cover glasses.
Certificates
Search by Lot NumberProduct Family Options
Product Information
- Depth of chamberAvailabilityPrice
- MARI0640130Counting chamber, bright-lined, Neubauer
- MARI0640110Counting chamber, dark-lined, Neubauer
- MARI0640111Counting chamber, dark-lined, with clamps
- MARI0640131Counting chamber, bright-lined, with clamps, NeubauerCounting chamber, bright-lined, with clamps, Neubauer0,10 mmLog in for availability1 * 1 ST131,00 €