Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Water

Glass bottle, 500 mL
Catalog # SERA39798.03
Marketsource
Supplier:   Serva
Water
Catalog # SERA39798.03
Supplier:  Serva
Marketsource
CAS Number:  7732-18-5
Water

Glass bottle, 500 mL

CAS Number:  7732-18-5
Price
39,00 €
Connexion Pour la disponibilité
1 * 500 mL
Product Details & Documents
Synonymes:  Hydrogen oxide
  • Numéro CAS:7732-18-5
  • Formule:H₂O
  • Numéro MDL:MFCD00011332
  • Poids moléculaire:18.02 g/mol
  • Index Merck:13,10098
  • Point d'ébullition:100 °C (1013 hPa)
  • Point de fusion:0 °C
  • Densité:1 g/cm³ (4 °C)
  • Température de stockage:Ambient
  • EINECS:231-791-2
Spécifications

Spécifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.