Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Water

Marketsource
Fournisseur:   Roth Carl
Water
Fournisseur:  Roth Carl
Marketsource
Certificates

5 Options Available Below

Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2
Product Family Options

Product Information

  • Pack type
    Pk
    Disponibilité
    Prix
  • Glass bottle
    1 mL
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - 122,50 €
    122,50 €
    1 * 1 mL
       122,50 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    20 mL
    Log in for availability
    Log in for availability
    1 * 20 mL
    1 * 20 mL - 32,50 €
    32,50 €
    1 * 20 mL
       32,50 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 30,50 €
    30,50 €
    1 * 250 mL
       30,50 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 38,50 €
    38,50 €
    1 * 500 mL
       38,50 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 52,90 €
    52,90 €
    1 * 1 L
       52,90 €

    Specifications:
    Marketsource
    Marketsource