Water
Marketsource
Supplier: Roth Carl
About this item
Synonyms: Hydrogen oxide
- CAS Number:7732-18-5
- Formula:H₂O
- MDL Number:MFCD00011332
- Molecular Weight:18.02 g/mol
- Merck Index:13,10098
- Boiling Pt:100 °C (1013 hPa)
- Melting Pt:0 °C
- Density:1 g/cm³ (4 °C)
- Storage Temperature:Ambient
- EINECS:231-791-2
2 Options Available Below
Product Details & Documents
Synonyms: Hydrogen oxide
- CAS Number:7732-18-5
- Formula:H₂O
- MDL Number:MFCD00011332
- Molecular Weight:18.02 g/mol
- Merck Index:13,10098
- Boiling Pt:100 °C (1013 hPa)
- Melting Pt:0 °C
- Density:1 g/cm³ (4 °C)
- Storage Temperature:Ambient
- EINECS:231-791-2
Documents
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
Specifications
- Catalog No:
- ROTHA511.1
- ROTHA511.3
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
- EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
Safer Human and Environmental Health MarketsourceMore
Specifications
Cat. No.ROTHA511.1Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationEPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussionSafer Human and Environmental Health Marketsource - Specifications:
Safer Human and Environmental Health MarketsourceMore
Specifications
Cat. No.ROTHA511.3Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationEPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussionSafer Human and Environmental Health Marketsource



