Orders LinkContactUsLinkComponent
Staining tiles
Staining tiles
Supplier:   MARIENFELD
Staining tiles
Supplier:  MARIENFELD
Certificates
Staining tiles
6 Options Available Below
Product Details & Documents
  • Bevelled edges
  • Ground and polished cavities
  • Clipped corners: Except the one with twelve wells
Product Family Options
Product Information
  • No. of wells
    Well size
    Colour
    W×D×H
    Availability
    Price
  • 12
    Ø×H: 16×1,5 mm
    Transparent
    76×60×4 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 1 680,00 €
    1 680,00 €
    1 * 10 ST
       1 680,00 €
  • 6
    Ø×H: 20/22×2 mm
    Transparent
    130×100×6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 360,00 €
    360,00 €
    1 * 1 ST
       360,00 €
  • 24
    Ø×H: 24×3 mm
    Transparent
    130×200×6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 563,00 €
    563,00 €
    1 * 1 ST
       563,00 €
  • 30
    Ø×H: 20/22×2 mm
    Transparent
    130×180×6 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST
  • 12
    Ø×H: 20/22×2 mm
    Transparent
    130×100×6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 396,00 €
    396,00 €
    1 * 1 ST
       396,00 €
  • 24
    Ø×H: 20/22×2 mm
    Transparent
    130×160×6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 468,00 €
    468,00 €
    1 * 1 ST
       468,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.