Order Entry
Belgium
Orders LinkContactUsLinkComponent
Water

Water

Marketsource
Supplier:   NEN LIFE SCIENCE
Water
Supplier:  NEN LIFE SCIENCE
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 1
    Log in for availability
    Log in for availability
    1 * 1 Un.
    1 * 1 Un.

    Specifications:
    Marketsource
    Marketsource
  • 1
    Log in for availability
    Log in for availability
    1 * 1 Un.
    1 * 1 Un.

    Specifications:
    Marketsource
    Marketsource