Orders LinkContactUsLinkComponent
Embedding cassettes
Embedding cassettes
Supplier:   MARIENFELD
Embedding cassettes
Supplier:  MARIENFELD
Certificates
Embedding cassettes
Embedding cassettes
2 Options Available Below
Product Details & Documents
Plastic (POM).
  • Resistant against the solvents usually used in laboratories
  • Different colours offer the possibility of colour coding (for example for different users, priorities)
  • One 45° inclined and two vertical marking areas
  • Roughness of the marking areas optimized to allow labelling by different systems
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • MARI5253104
    Embedding cassettes
    Embedding cassettes
    White
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 116,00 €
    116,00 €
    1 * 500 ST
       116,00 €
  • MARI5253109
    Embedding cassettes
    Embedding cassettes
    Orange
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 116,00 €
    116,00 €
    1 * 500 ST
       116,00 €