Order Entry
Belgium
ContactUsLinkComponent
Water, Ultrapur for trace analysis, Supelco®
Water, Ultrapur for trace analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Water, Ultrapur for trace analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Molar Mass:
    18.02 g/mol
  • Boiling Point:
    100 °C (1013 hPa)
  • Vapor Pressure:
    23 hPa (20 °C)
  • Density:
    1.00 g/cm3 (20 °C)
  • pH:
    (H2O, 20 °C) neutral

Specification Test Results

Related Information

Frequently Bought Together